missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Stainless Steel Wire Brush

Durable stainless steel wire brushes with nylon bristles are available in a variety of sizes for cleaning parts, tubing and hard to reach areas.

Supplier:  Micronova™ BRWM22512J

 View more versions of this product

Catalog No. 19435118


Only null left

Description

Description

Micronova Stainless Steel Wire Brushes are excellent for cleaning valves, tubing, cylinders, narrow chambers and other hard to clean areas. The durable brushes are flexible enough bend through fittings but stiff enough for removing residues. The tapered versions add a level of cleaning versatility.

  • Nylon bristle brushes with a stainless steel wire core
  • Ideal for valves, tubing, cylinders and narrow chambers
  • Flexible to bend through fittings but stiff enough to remove residues
  • Tapered versions add cleaning versatility
  • Available in a range of diameters from 0.25 to 2 inches
Specifications

Chemical Identifiers

Nylon
ISO 9001 Certified Facility; Certificate of Conformance; FDA Approved Materials
Reusable
No
Standard disinfectants and cleaning solutions
Non-sterile
Store in a dry area, at ambient temperature with ideal air flow.
Non-filtered
Each brush is individually packaged

Specifications

J-Shaped Brush
Medium Stiffness
Stainless Steel Wire with Plastic Grip
Tuft End
Wire brush^ Flexbrush, flexible inchjinch shaped, stainless steel wire brush on plastic handle with acme connection.
Item can be cleaned after use and hung up for optimal drying and sanitation purposes. Flexibility of the head allows users to 'shape' the head for custom cleaning applications
White
2.25 in.
16 x 2.2 x 2.2 in. (40.6 x 5.7 x 5.7 cm)
Non-filtered
Brushes flex to clean ducting, piping exteriors or vessel interiors.
Without Handle
12 in.
40.6 cm
10.2 cm
Each brush is individually packaged
Standard disinfectants and cleaning solutions
9 lb.
2.25 in.
Nylon
Yes
Acme Thread
Non-sterile
Autoclavable
ISO 9001 Certified Facility; Certificate of Conformance; FDA Approved Materials
Store in a dry area, at ambient temperature with ideal air flow.
5.72 cm
Reusable
No
STAH or STAEH series handles; FlexBrushTM Covers
16 in.
4 in.
30.4 cm
Nylon, Stainless Steel (304), Plastic
12/Cs.
16 x 10 x 8 in.
4.08 kg
5.7 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions