missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel wire, 0.38mm (0.015in) dia, annealed, Type 304

Catalog No. AA43301BY
Change view
Click to view available options
Quantity:
100 cm
500 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43301BY 100 cm
AA43301CC 500 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43301BY Supplier Thermo Scientific Chemicals Supplier No. 043301BY

Temper: soft | Fe:Cr:Ni; 70:19:11 wt%

Stainless Steel Wire are suitable for making Kitchenware, Basket, Cable Trays and Trolleys with electro-polishing as the final process . Clean, de-greased & defect free surface results in very lustrous appearance when electro polished.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Color Silver
Chemical Name or Material Stainless Steel
Name Note annealed, Type 304
Appearance Silver-colored wire
Quantity 100 cm
Odor Odorless
Form Wire
Molecular Formula Fe:Cr:Ni; 70:19:11 wt%
MDL Number MFCD00148499
Solubility Information Insoluble in water.

RUO – Research Use Only