Learn More
Thermo Scientific™ Stainless Steel Union, compatible with Upchurch and Parker Fittings, 0.010 in. through hole, 0.025µL swept volume DFS Item
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Supplier:  Thermo Scientific™ UPU435
Catalog No. 29-008-261
Only null left
Explore available promotions
Description
Description
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
  • Absolute zero or low dead volume formats
  • Includes two stainless steel nuts and ferrules
Product Suggestions
Product Suggestions