Learn More
Millipore Canada Ltd Swab Rack DFS Item
Supplier:  MilliporeSigma 1465300001
Swab rack is used to hold the ICR Swabs that are designed for environmental monitoring of dry surfaces in cleanrooms and isolators. The swabs after sampling can be transported and incubated in an upright position using the Stainless-steel swab racks that can hold upto 40 ICR-Swabs.
Catalog No. NC0278264
Only null left
Explore available promotions