Learn More
Stainless Steel Spheres, 4mm (0.16 in.) dia., Thermo Scientific Chemicals
$174.85 - $174.85
Specifications
| Chemical Name or Material | Stainless Steel |
|---|---|
| Appearance | Spheres |
| Form | Sphere |
| Molecular Formula | Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt% |
| MDL Number | MFCD00148499 |
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Stainless Steel | |
| Sphere | |
| MFCD00148499 |
| Spheres | |
| Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt% | |
| Insoluble in water. |
RUO – Research Use Only