Learn More
Bel-Art™ Stainless-steel Sampling Spoon and Spatula
Description
Ends coated with brown Fluo-Kem™ PTFE FEP.
- Coating provides anti-stick, corrosion-resistant surface
- Rod flattened at one end to serve as spatula with spoon on opposite end
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Product Type | Spoon and Spatula Sampler |
| Finish | Brown Fluo-Kem PTFE FEP |
| Material | Stainless Steel |
| Length (Metric) | 22.9 cm |