Learn More
Anton Paar STAINLESS STEEL, LUER ADAPTER, FOR PELTIER, 2.5mm for MCP 150
Catalog No. NC1818320
Encompass
2.5 mm stainless steel sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to the MCP 150.Filling volume approximately 0.05 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 150 item NC1779945
Catalog No. NC1818320 Supplier Anton Paar Supplier No. ANP 109524
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
2.5 mm stainless steel sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to the MCP 150.Filling volume approximately 0.05 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 150 item NC1779945