missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Stainless Steel Quick Link

Catalog No. 14727065
Encompass
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14727065 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14727065 Supplier Bio-Serv™ Supplier No. BIO9133
  • Made of stainless steel
  • Use to suspend various enrichment devices
  • Autoclavable
  • Cagewasher safe
  • 1.75 in. long x 3/8 in. inside width x 1/8 in. thick with a 3/8 in. opening

Specifications

Autoclavable Autoclavable
Color Silver
Material Stainless Steel
For Use With (Equipment) 3/8in. opening
Thickness (English) Wall 1/8 in.
Length (English) 1 3/4 in.
Width (English) 3/8 in.