Learn More
Bal Supply Stainless Steel Laboratory Scoop
Catalog No. 13684166
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
13684166 Stainless Steel
1 options
Catalog No. 13684166 Supplier Bal Supply Supplier No. LS65SS
Bal Supply Stainless Steel Laboratory Scoop has one pointed, and one blunt end
  • Stainless Steel
  • 6.5 inches long
  • Will not rust
  • One end is blunt, the other is pointed
Specifications
Shape One end is blunt, the other is pointed
Sterility Non-sterile
Autoclavable Autoclavable
Color Silver
Disposable Single-use
Product Type Scoop
Length (English) 6.5 in.
Dimensions (L x W) 6.5 x 0.5 x 0.5 in.
Material Stainless Steel