missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Stainless Steel Micro Spatula Reagent Scoop

Catalog No. S41700
Change view
Click to view available options
Length (Metric):
150 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S41700 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S41700 Supplier Eisco™ Supplier No. CH0635D
Only null left

Stainless steel micro spatula features a scoop on one end and flat edge on the other end.

  • Scoop on one end and flat edge on the other end
  • Ideal for adding small amounts of ingredients and mixing
  • Autoclavable and chemical resistant
  • Single Scoop

Warranty

1 year limited manufacturer warranty

Specifications

Quantity 1
Shape Scoop End, Spatula End
Autoclavable Autoclavable
Color Silver
Product Type Double Ended Spatula
For Use With (Application) Measuring
Length (English) 6 in.
Diameter (English) Outer Handle: 0.12 in.
Diameter (Metric) Outer Handle: 3 mm
Dimensions (L x W) Scoop: 0.51 x 0.20 in.; Flat Side: 1.96 x 0.27 in.
Material Stainless Steel
Length (Metric) 150 mm
Show More Show Less