missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Stainless Steel Hooked Weights Set

Catalog No. S88341
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88341 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88341 Supplier Eisco™ Supplier No. PH0037DSSECO

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stainless Steel Hooked Weights Set feature top and bottom retractable hooks (except the 10 g weight) to allow pieces to be hooked in tandem or sit flat on a surface.

  • Weights range from 10-1000 grams: (1) 10g, (2) 20g, (1) 50g, (1) 100g, (2) 200g, (1) 500g, (1) 1000g
  • Includes a polished wooden storage block (8 x 3.75 x 3 in.)
  • Entire set weighs approx. 6.5 lbs

Warranty

1 year limited manufacturer warranty

Specifications

Product Type Hook Weight Set
Material Stainless Steel, Wood
Weight (Metric) 2.9 kg
Length (Metric) 20.3 cm
For Use With (Application) Calibration
Includes 9 weights, Wooden block
Quantity 1 Set