Learn More
Medegen Stainless-Steel Graduates
Graduated inside in oz. and mL
$113.39 - $113.39
Specifications
Certifications/Compliance NSA
Material Stainless Steel
Autoclavable Autoclavable
Product Type Pharmaceutical Graduated Cylinder
Type Pharmaceutical Graduate
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (English) Height (English) Diameter (English) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Height (English) Diameter (English) Price Quantity  
11-420-10B
View Documents
Medegen
906830
32 oz. 4.62 in. 5.25 in.
Each for $113.39
Case of 6 Each for $680.34
Only null left
Description
Description
  • Convenient handle
  • Smooth-pour spout
Specifications
Specifications
NSA
Autoclavable
Pharmaceutical Graduate
Stainless Steel
Pharmaceutical Graduated Cylinder