missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel Foil, 0.2mm (0.008 in.) thick, Thermo Scientific Chemicals

$105.58 - $105.58

Specifications

Chemical Name or Material Stainless Steel
Color Silver
Name Note Type 304
Appearance Silver-colored foil
Odor Odorless
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42255GH
View Documents
Thermo Scientific Chemicals
042255GH
100 x 100 mm
Each for $105.58
Only null left
 
Description

Description

It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Stainless Steel
Type 304
Odorless
Fe:Cr:Ni; 70:19:11 wt%
Insoluble in water.
Silver
Silver-colored foil
Foil
MFCD00148499
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only