missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel foil, 0.1mm (0.004in) thick, Type 304

Catalog No. AA41583FF
Change view
Click to view available options
Quantity:
25 x 25 mm
100 x 100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41583FF 25 x 25 mm
AA41583GH 100 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41583FF Supplier Thermo Scientific Chemicals Supplier No. 041583FF
Only null left

Fe:Cr:Ni; 70:19:11 wt%

It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Color Silver
Chemical Name or Material Stainless Steel
Appearance Silver -colored foil
Quantity 25 x 25 mm
Odor Odorless
Form Foil
Molecular Formula Fe:Cr:Ni; 70:19:11 wt%
MDL Number MFCD00148499
Solubility Information It is insoluble in water.

RUO – Research Use Only