Learn More
Elma™ Stainless Steel Baskets for Elmasonic Xtra ST Ultrasonic Cleaner
For use with Elmasonic Xtra ST Ultrasonic Cleaner.
Supplier:  Elma™ 102 6584
 View more versions of this product
Catalog No. 08000109
Only null left
Description
Description
  • Holds parts when immersing and removing from solution
  • Open weave in sides and bottoms
  • Protect cleaner and parts from damage caused by direct contact
Specifications
Chemical Identifiers
Basket
Hold Parts When Immersing and Removing Them From Solution in Ultrasonic Cleaners
Stainless Steel
Specifications
Basket
Hold Parts When Immersing and Removing Them From Solution in Ultrasonic Cleaners
39.8 x 25.4 x 21.8 cm
Ultrasonic Cleaner, Elmasonic Xtra ST 600 H
Stainless Steel
Product Suggestions
Product Suggestions