Learn More
Elma™ Stainless Steel Baskets for Elmasonic Xtra ST Ultrasonic Cleaner
Catalog No. 08000108
Change view
Click to view available options
For Use With (Equipment):
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H
Ultrasonic Cleaner, Elmasonic Xtra ST 1600 H
Ultrasonic Cleaner, Elmasonic Xtra ST 500 H
Ultrasonic Cleaner, Elmasonic Xtra ST 600 H
Ultrasonic Cleaner, Elmasonic Xtra ST 800 H
Dimensions (L x W x H):
24.5 x 22.8 x 37 cm
39.8 x 25.4 x 21.8 cm
39.8 x 25.4 x 37 cm
48 x 52.5 x 29.9 cm
49 x 52.5 x 20.8 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Dimensions (L x W x H)
08000108 Ultrasonic Cleaner, Elmasonic Xtra ST 500 H 24.5 x 22.8 x 37 cm
08000109 Ultrasonic Cleaner, Elmasonic Xtra ST 600 H 39.8 x 25.4 x 21.8 cm
08000112 Ultrasonic Cleaner, Elmasonic Xtra ST 1600 H 48 x 52.5 x 29.9 cm
08000110 Ultrasonic Cleaner, Elmasonic Xtra ST 800 H 39.8 x 25.4 x 37 cm
08000111 Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H 49 x 52.5 x 20.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 08000108 Supplier Elma™ Supplier No. 107 6253
Only null left
For use with Elmasonic Xtra ST Ultrasonic Cleaner.
  • Holds parts when immersing and removing from solution
  • Open weave in sides and bottoms
  • Protect cleaner and parts from damage caused by direct contact
Specifications
Type Basket
For Use With (Equipment) Ultrasonic Cleaner, Elmasonic Xtra ST 500 H
For Use With (Application) Hold Parts When Immersing and Removing Them From Solution in Ultrasonic Cleaners
Material Stainless Steel
Dimensions (L x W x H) 24.5 x 22.8 x 37 cm