Learn More
GA International Stain-Proof Xylene-Resistant Labels 0.866"" x 0.59"" / 22mm x 15mm Rectangle White From -80C to +149C (-112F to +300F)
Catalog No. 5000570883
Encompass
Thermal-transfer printable labels designed for microscope slides. These highly glossy labels repel stains and do not acquire colors from histological stains, such as hematoxylin or eosin. The labels and printed information are xylene-resistant when used in combination with our XAR-class of solvent-resistant ribbons. The print will resist xylene, toluene, alcohol, MEK, and other solvents and chemicals without needing additional protection. Ideal for histology lab applications.
Catalog No. 5000570883 Supplier GA International Supplier No. XST117NPC33WH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermal-transfer printable labels designed for microscope slides. These highly glossy labels repel stains and do not acquire colors from histological stains, such as hematoxylin or eosin. The labels and printed information are xylene-resistant when used in combination with our XAR-class of solvent-resistant ribbons. The print will resist xylene, toluene, alcohol, MEK, and other solvents and chemicals without needing additional protection. Ideal for histology lab applications.