missing translation for 'onlineSavingsMsg'
Learn More

BD Stain Droppers

Catalog No. 1491017
Change view
Click to view available options
Stain Type:
Dobell and O'Connor's Iodine
Lactophenol Cotton Blue
Methylene Blue
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stain Type
1491017 Dobell and O'Connor's Iodine
1491015 Lactophenol Cotton Blue
1491076 Methylene Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1491017 Supplier BD Supplier No. 261189

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify microorganisms without mixing or measuring stains

To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.

  • Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
  • Strict quality controls assure uniformity and accuracy

Specifications

Quantity 50 Pack
Certifications/Compliance CE
Format Dropper
For Use With (Application) Used in the staining of cysts and eggs of parasites found in human feces
Storage Requirements 15° to 30°C
Stain Type Dobell and O'Connor's Iodine
Volume (Metric) 0.5 mL
Product Type Reagent/Stain Dropper