Learn More
Fisherbrand™ Stage Micrometer Eyepiece Reticle
Description
- 1 mm long
- 100 divisions
- Stage micrometer
- 10 points NIST standard
Specifications
Specifications
| Type | Microscope Eyepiece Reticle |
| For Use With (Equipment) | Microscope |