Learn More
BINDER™ Stacking Adapter
Description
- Used to safely stack two identical devices
- For use with KB 65, KB 130, KB PRO 130, KBF 130, or KBF PRO 130 models
Specifications
Specifications
| For Use With (Equipment) | KB 65 |
| Material | Plastic |
| Type | Stacking Adapter |