Learn More
Analytical Instruments Group (Aig) Stable-Store2 Buffer, 1 Liter Size
Catalog No. 501351693
Encompass
Stable-Store2, 1 Liter Size. Stable-Store in combination with Stable-Lyse is used to preserve whole blood and cell suspensions.
Catalog No. 501351693 Supplier Analytical Instruments Group (Aig) Supplier No. STBLSTORE21000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stable-Store2, 1 Liter Size. Stable-Store in combination with Stable-Lyse is used to preserve whole blood and cell suspensions.