Learn More
BeLLCo Glass Steel 2-Port Assembly 2L For Media Bottle
Catalog No. 501945776
Encompass
These stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media or liquids. The 2- port models feature a siphon vent tube and is supplied with a 45mm polypropylene blue open top screw cap and white silicone gasket. Specifications: Designed For 5637 And 5639 Media Storage Bottles.Welded, 316l Stainless Steel Construction 1/4 Inch Od Tubing Ra32 Finish Or Better.Electropolished Finish, Continuous Seal Weld.Two Port Model Features Siphon And Vent Tubes.Supply W/45mm S/Cap & White Silicone Liner.Long Tube: 8 1/2.
Catalog No. 501945776 Supplier BeLLCo Glass Supplier No. 563720002R
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media or liquids. The 2- port models feature a siphon vent tube and is supplied with a 45mm polypropylene blue open top screw cap and white silicone gasket. Specifications: Designed For 5637 And 5639 Media Storage Bottles.Welded, 316l Stainless Steel Construction 1/4 Inch Od Tubing Ra32 Finish Or Better.Electropolished Finish, Continuous Seal Weld.Two Port Model Features Siphon And Vent Tubes.Supply W/45mm S/Cap & White Silicone Liner.Long Tube: 8 1/2.