Learn More
Chemglass Inc Dish Only, 150 x 75mm, Oil Bath, Stainless Steel
Catalog No. CG1100D02
Encompass
Dish Only, 150 x 75mm, Oil Bath, Stainless Steel. Stainless steel dish with lid for use as an oil bath when used in conjunction with a hot plate stirrer having a thermocouple connection. Available in three sizes, complete with lids.
Catalog No. CG1100D02 Supplier Chemglass Inc Supplier No. CG1100D02
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Dish Only, 150 x 75mm, Oil Bath, Stainless Steel. Stainless steel dish with lid for use as an oil bath when used in conjunction with a hot plate stirrer having a thermocouple connection. Available in three sizes, complete with lids.