missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Chloride, Crystal, 99%, Spectrum™ Chemical

Catalog No. 18610569
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610569 50 kg Amber Glass Bottle
18610564 25 g Amber Glass Bottle
18610565 125 g Amber Glass Bottle
18610568 500 g Amber Glass Bottle
18610567 2.5 kg Amber Glass Bottle
18610566 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610569 Supplier Spectrum Chemical Mfg Cor Supplier No. F105550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2FeH8O4
Molecular Weight (g/mol) 198.81
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
IUPAC Name tetrahydroxyironbis(ylium) dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Fe++]

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 50 kg
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Ferrous Chloride, Crystal,

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.