Learn More
Biosan Laboratories Inc SRB KIT 25 TEST/BX
Supplier: Biosan Laboratories Incorporated BIO100
Sani-Check SRB Kit; 25 Tests; Contains tubes of culture media specifically formulated to promote the growth of anaerobic sulfate reducing bacteria; Used in areas where little or no formal laboratory facilities are available; 1/KT