Learn More
Abcam Inc SR-9009, REV-ERBalpha/beta agonist, 5MG
Catalog No. 502704887
Encompass
MW 437.94 g/mol. Potent REV-ERB/ agonist (IC values are 670 and 800 nM for Rev-ErbA and Rev-ErbA-dependent repressor activity respectively). Modulates circadian behavior, shows antiobesogenic and anti-inflammatory effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502704887 Supplier Abcam Inc Supplier No. AB1461745MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 437.94 g/mol. Potent REV-ERB/ agonist (IC values are 670 and 800 nM for Rev-ErbA and Rev-ErbA-dependent repressor activity respectively). Modulates circadian behavior, shows antiobesogenic and anti-inflammatory effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement