missing translation for 'onlineSavingsMsg'
Learn More

Sqwincher™ Activity Drinks, Fast Pak Liquid Concentrate

Catalog No. 19043426
Change view
Click to view available options
Flavor:
Fruit Punch
Lemonade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor
19043426 Fruit Punch
19043431 Lemonade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19043426 Supplier Sqwincher™ Supplier No. 11081

Thirst-quenching drink that's low in sodium and high in potassium

  • Formulated with electrolytes and nutrients
  • Replenishes fluids, minerals, and sugars depleted through physical exertion
  • Prevents or reduces severity of heat-stress disorders
  • Provides supplemental source of energy
  • Just add water to package — it's your cup
  • Makes 6 fl. oz.

Specifications

Product Type Activity Drink
Flavor Fruit Punch
Format Fast Pack Liquid Concentrate
For Use With (Application) Provides supplemental source of energy, prevents or reduces severity of heat-stress disorders
Quantity 200 Cs.
Volume (English) 0.6 oz.
Volume (Metric) 18 mL