missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Squids™ Tool Tethering Kit
Description
- Straightforward installation
- Lack of connection points for tool lanyards can create a hazardous environment while working at heights
- Measure and cut tape to the specified length listed in instructions. Lay the tool tail on the tool being tethered and wrap the tape once around itself beginning below one of the web catch points. Continue wrapping and stretching the tape firmly. Firmly push down on the end and attach the Squids™ Tool Lanyard to complete the tethering system.
Specifications
Specifications
| Includes | Squids™ 3100F(x) Single Carabiner Tool Lanyard, a 12 ft / 3.7 m roll of Squids™ 3755 Self-Adhering Tape Trap, and three Squids™ 3713 Swiveling Elastic Tool Tails. |
| Certifications/Compliance | ANSI/ISEA 121-2018 |
| Load Bearing Capacity (English) | Up to 3 x 10 lb. tools |
| Load Bearing Capacity (Metric) | Up to 3 4.5 kg tools |
| For Use With (Equipment) | Claw hammers, sledgehammers, mallets, pliers, bolt cutters, spud wrenches and bull pins |
| Size | Up to 3 tools |