missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Squids™ Tool Tethering Kit
Description
- Straightforward installation
- Lack of connection points for tool lanyards can create a hazardous environment while working at heights
- Measure and cut tape to the specified length listed in instructions. Lay the tool tail on the tool being tethered and wrap the tape once around itself beginning below one of the web catch points. Continue wrapping and stretching the tape firmly. Firmly push down on the end and attach the Squids™ Tool Lanyard to complete the tethering system.
Specifications
Specifications
| Certifications/Compliance | ANSI/ISEA 121-2018 |
| For Use With (Equipment) | Claw hammers, sledgehammers, pliers, bolt cutters, spud wrenches, bull pins, levels and pry bars |
| Load Bearing Capacity (English) | Up to 4 x 5 lb. tools |
| Includes | Squids™ 3102F(x) Detachable Single Carabiner Lanyard, a 12 ft / 3.7 m roll of Squids™ 3755 Self-Adhering Tape Trap, and three Squids™ 3103 Detachable Elastic Tool Tails. |
| Load Bearing Capacity (Metric) | Up to 4 2.3 kg tools |
| Size | Up to 4 tools |