missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Squids™ 3158 Coil Tool Lanyard with Clamp

Catalog No. 19910454
Change view
Click to view available options
Color:
Black
Lime
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19910454 Black
19910455 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19910454 Supplier Ergodyne™ Supplier No. 19158

Squids™ 3158 Coil Tool Lanyard with Clamp is compact and used to tether small hand tools, which prevents falling objects when working at heights.

  • Low-profile durable plastic coil to withstand even the toughest conditions
  • Easily attaches to hard hats and other small hand tools for safekeeping
  • Coiled lanyard design prevents snag and tangle hazards
  • Quick connect clamp attaches securely to fabric
  • Tethering loop to securely connect to an anchor or tool attachment
  • Certified to a 2:1 safety factor

Specifications

Color Black
Length (English) 12 to 48 in.
Length (Metric) 30 to 122 cm
Load Bearing Capacity (English) 2 lb.
Load Bearing Capacity (Metric) 0.9 kg