missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Squids™ 3157 Coil Tool Lanyard with Buckle

Catalog No. 19910453
Change view
Click to view available options
Color:
Black
Lime
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19910453 Lime
19910452 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19910453 Supplier Ergodyne™ Supplier No. 19157

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Squids™ 3157 Coil Tool Lanyard with Buckle is compact and used to tether small hand tools, which prevents falling objects when working at heights.

  • Approved to the ANSI/ISEA 121-2018 Standard
  • Low-profile durable plastic coil to withstand even the toughest conditions
  • Easily attaches to hard hats and other small hand tools for safekeeping
  • Coiled lanyard design prevents snag and tangle hazards
  • Double lock buckle attaches to harness
  • Tethering loop to securely connect to an anchor or tool attachment
  • Certified to a 2:1 safety factor

Specifications

Color Lime
Load Bearing Capacity (English) 2 lb.
Load Bearing Capacity (Metric) 0.9 kg
Length (Metric) 30 to 122 cm
Certifications/Compliance Approved to the ANSI/ISEA 121-2018 standard
Length (English) 12 to 48 in.