missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Squids™ 3157 Coil Tool Lanyard with Buckle
Description
- Approved to the ANSI/ISEA 121-2018 Standard
- Low-profile durable plastic coil to withstand even the toughest conditions
- Easily attaches to hard hats and other small hand tools for safekeeping
- Coiled lanyard design prevents snag and tangle hazards
- Double lock buckle attaches to harness
- Tethering loop to securely connect to an anchor or tool attachment
- Certified to a 2:1 safety factor
Specifications
Specifications
| Color | Black |
| Length (English) | 12 to 48 in. |
| Length (Metric) | 30 to 122 cm |
| Certifications/Compliance | Approved to the ANSI/ISEA 121-2018 standard |
| Load Bearing Capacity (English) | 2 lb. |
| Load Bearing Capacity (Metric) | 0.9 kg |