missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Squids™ 3130M Coiled Cable Lanyard

Catalog No. 19820387
Change view
Click to view available options
Load Bearing Capacity (Metric):
2.26 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity (Metric)
19820387 2.26 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820387 Supplier Ergodyne™ Supplier No. 19131

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Squids™ 3130M Coiled Cable Lanyard reduces snag hazards of traditional lanyards while preventing dropped objects.

  • Approved to the ANSI/ISEA 121-2018 standard
  • Constructed with PU covered aircraft cable
  • Stainless steel carabiners
  • Coil design prevents snag and tangle hazards
  • Swiveling design prevents tangles and fast recoil
  • Captive eye carabiner
  • Certified to a 2:1 safety factor

Specifications

Color Orange
Material Stainless Steel
Length (English) 6.5 to 48 in.
Certifications/Compliance ANSI/ISEA 121-2018 standard
Load Bearing Capacity (English) 5 lb.
Length (Metric) 16.5 to 123 cm
Load Bearing Capacity (Metric) 2.26 kg