missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Squids™ 3116 Pull-On Wrist Lanyard with Buckle

Catalog No. 19820385
Change view
Click to view available options
Load Bearing Capacity (Metric):
1.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity (Metric)
19820385 1.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820385 Supplier Ergodyne™ Supplier No. 19057

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Squids™ 3116 Pull-On Wrist Lanyard with Buckle is designed to tether tools and reduces snag hazards.

  • Elastic maintains snug fit
  • Pull-on design is easy to take on and off
  • One size fits all
  • Modular quick connecting buckle for exchanging multiple tools
  • Compatible with 3103: Detachable Lanyard Loops
  • Tested using a 2:1 safety factor

Specifications

Color Black
Load Bearing Capacity (English) 3 lb.
Load Bearing Capacity (Metric) 1.5 kg
Certifications/Compliance ANSI/ISEA 121-2018 standard