missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Squids™ 3115 Wrist Tool Lanyard

Catalog No. 19820383
Change view
Click to view available options
Product Type:
Large/X-Large Lanyard
Small/Medium Lanyard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19820383 Large/X-Large Lanyard
19820382 Small/Medium Lanyard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19820383 Supplier Ergodyne™ Supplier No. 19052

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Adjustable wrist lanyard tethers tools, which limits the distance of a dropped tool and reduces snag hazards.

  • Approved to the ANSI/ISEA 121-2018 standard
  • Polyester webbing cuff secures to wrist
  • Energy absorbing ribbed pull cord
  • Adjustable loop with barrel lock for tool attachment
  • Short length improves the drop clearence needed
  • Tether length: 7.5 in. (19 cm)
  • Maximum safe working capacity: 2 lb. (0.9 kg)
  • Certified to a 2:1 safety factor

Specifications

Color Black
Certifications/Compliance Approved to the ANSI/ISEA 121-2018 standard
Length (English) 5 to 7 in.
Product Type Large/X-Large Lanyard
Length (Metric) 13 to 18 cm