missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Squids™ 3001 Retractable Tool Lanyard with Carabiner and Loop Attachments

Catalog No. 19820314
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820314 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820314 Supplier Ergodyne™ Supplier No. 19301
Only null left

Squids™ 3001 Retractable Tool Lanyard with Carabiner and Loop Attachments is designed to mount to the users belt or a separate anchor point.

  • Retractable design limits snags, tangles and tripping hazards
  • Modular connecting buckle to quickly exchange tools when used together with the Squids™ 3026 Loop Attachments Accessory 3-Pack
  • Swiveling ring for 360 degree rotation works well with rotating tools to reduce tangles
  • Self-setting pawls set desired lanyard length to take the tension out
  • ABS thermoplastic housing protects the internal components and Technora cord for ultimate strength and durability

Specifications

Load Bearing Capacity (Metric) 0.9 kg
Size Standard
Load Bearing Capacity (English) 2 lb.