Learn More
Corning™ Square and Rectangular Cover Glasses
GREENER_CHOICE
Provides quality, reliability and performance for your laboratory microscopy needs
Supplier:  Corning™ 285522
 View more versions of this product
Catalog No. 12553457
Only null left
Description
Description
Provides quality, reliability and performance for your laboratory microscopy needs
  • Made from soda-lime glass and borosilicate glass with high clarity
  • Available in square and rectangular shapes, and in three standard sizes and thicknesses
Order Info
Packaged in plastic boxes for protection during storage.
Specifications
Chemical Identifiers
Coverslip
Borosilicate Glass
Non-sterile
Microscopes
Specifications
Coverslip
Borosilicate Glass
22 mm
Non-sterile
100/Pk.
Square
22 mm
0.17 to 0.25 mm
Microscopes
Product Suggestions
Product Suggestions