missing translation for 'onlineSavingsMsg'
Learn More

Squalene 98.0+%, TCI America™

Catalog No. H0097500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0097500ML 500 mL
H009725ML 25 mL
H0097100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0097500ML Supplier TCI America Supplier No. H0097500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-02-4
Molecular Formula C30H50
Molecular Weight (g/mol) 410.73
MDL Number MFCD00008912
InChI Key YYGNTYWPHWGJRM-AAJYLUCBSA-N
Synonym squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
PubChem CID 638072
ChEBI CHEBI:15440
IUPAC Name (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 252°C
Quantity 500 mL
Formula Weight 410.73
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Squalene
EINECSNumber (9)-0761
RTECSNumber KB6010000
TSCA Yes