missing translation for 'onlineSavingsMsg'
Learn More

Squalene 98.0+%, TCI America™

Supplier:  TCI America H0097100ML

Quantity 100 mL

 View more versions of this product

Catalog No. H0097100ML


Only null left

Description

Description

Specifications

Chemical Identifiers

111-02-4
410.73
YYGNTYWPHWGJRM-AAJYLUCBSA-N
638072
(6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
C30H50
MFCD00008912
squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
CHEBI:15440
CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

Specifications

Squalene
-5°C
252°C
100 mL
squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C
410.73
CHEBI:15440
≥98.0% (GC)
111-02-4
Yellow
C30H50
MFCD00008912
YYGNTYWPHWGJRM-AAJYLUCBSA-N
(6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
638072
410.73
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0761

RTECSNumber : KB6010000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions