missing translation for 'onlineSavingsMsg'
Learn More

Squalene, MP Biomedicals™

Catalog No. ICN10294891
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10294891 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10294891 Supplier MP Biomedicals Inc Supplier No. 0210294891

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Functions as the biochemical precursor to the whole family of steroids. Squalene, MP Biomedicals is an isoprenoid compound that is structurally similar to beta-carotene and is an intermediate metabolite in the synthesis of cholesterol. Squalene is not ver

  • Functions in the skin as a quencher of singlet oxygen, protecting human skin surface from lipid peroxidation due to exposure to UV and other sources of ionizing radiation
  • Supplementation in mice has resulted in marked increases in cellular and non-specific immune functions in a dose-dependent manner
  • May act as a “sink” for highly lipophilic xenobiotics; since it is a nonpolar substance, it has a higher affinity for un-ionized drugs
  • Supplementation can reduce cholesterol and triglyceride levels; it might be a useful addition to potentiate the effects of some cholesterol-lowering drugs
  • The primary therapeutic use of squalene currently is as an adjunctive therapy in a variety of cancers

Chemical Identifiers

CAS 111-02-4
Molecular Formula C30H50
Molecular Weight (g/mol) 410.73
InChI Key YYGNTYWPHWGJRM-AAJYLUCBSA-N
Synonym squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
PubChem CID 638072
ChEBI CHEBI:15440
IUPAC Name (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

Specifications

Melting Point -75°C (literature)
Density 0.858g/mL at 25°C (Lit.)
Boiling Point 285°C (25mm (literature))
Refractive Index n20/D 1.494 (Lit.)
Quantity 1 L
Formula Weight 410.7
Percent Purity ≥98%
Physical Form Liquid
Recommended Storage Room Temperature (15–30°C).