missing translation for 'onlineSavingsMsg'
Learn More

Squalene, 99%, Spectrum™ Chemical

Catalog No. 18614005
Change view
Click to view available options
Quantity:
100 mL
500 mL
5 gal.
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614005 500 mL Amber Glass Bottle
18614002 100 mL Amber Glass Bottle
18614003 200 L Amber Glass Bottle
18614004 5 gal. Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614005 Supplier Spectrum Chemical Mfg Cor Supplier No. S2002500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-02-4

Specifications

Acidity 0.5
CAS Max % 1
Boiling Point 285°C
Packaging Amber Glass Bottle
Quantity 500 mL
Saponification Value 5
Percent Purity 0.99
Grade Reagent
Physical Form Liquid
Chemical Name or Material Squalene

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.