missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Squalene, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor S20025GLBL
| Quantity | 5 gal. |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 111-02-4 |
Specifications
| Squalene | |
| 111-02-4 | |
| 285°C | |
| 5 gal. | |
| 0.99 |
| 0.5 | |
| 1 | |
| Amber Glass Bottle | |
| 5 | |
| Liquid |