missing translation for 'onlineSavingsMsg'
Learn More

Squalene, 98%, Thermo Scientific Chemicals

Catalog No. AAB2094436
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2094436 500 g
AAB2094422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2094436 Supplier Thermo Scientific Chemicals Supplier No. B2094436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.

Solubility
Miscible with diethyl ether and acetone. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 111-02-4
Molecular Formula C30H50
Molecular Weight (g/mol) 410.73
MDL Number MFCD00008912
InChI Key YYGNTYWPHWGJRM-AAJYLUCBSA-N
Synonym squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
PubChem CID 638072
ChEBI CHEBI:15440
IUPAC Name (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

Specifications

Melting Point -75°C
Density 0.855
Boiling Point 285°C (25 mmHg)
Flash Point 218°C (424°F)
Assay Percent Range 0.97
Refractive Index 1.4955
Quantity 500 g
Sensitivity Air and light sensitive
Merck Index 14,8768
Solubility Information Miscible with diethyl ether and acetone. Immiscible with water.
Formula Weight 410.73
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Squalene
Show More Show Less
Hazard Category H304
Precautionary Statement P301+P310-P331-P501c
EINECSNumber 203-826-1
RTECSNumber XB6010000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only