missing translation for 'onlineSavingsMsg'
Learn More

Squalene, >98%, MP Biomedicals™

Catalog No. ICN10294890
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10294890 500 mL
ICN10294880 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10294890 Supplier MP Biomedicals Inc Supplier No. 0210294890

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-02-4
Molecular Formula C30H50
Molecular Weight (g/mol) 410.73
InChI Key YYGNTYWPHWGJRM-AAJYLUCBSA-N
Synonym squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
PubChem CID 638072
ChEBI CHEBI:15440
IUPAC Name (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

Specifications

Color Colorless
Density 0.855g/mL
Quantity 500 mL
Formula Weight 410.7
Physical Form Liquid
Chemical Name or Material Squalene