missing translation for 'onlineSavingsMsg'
Learn More

Squalane, ≥99.0% (GC), Solstice

Catalog No. 60047697
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60047697 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60047697 Supplier Solstice Supplier No. 8562950ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for GC,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 111-01-3
Molecular Formula C30H62
Molecular Weight (g/mol) 422.826
MDL Number MFCD00008953
InChI Key PRAKJMSDJKAYCZ-UHFFFAOYSA-N
Synonym squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane
PubChem CID 8089
IUPAC Name 2,6,10,15,19,23-hexamethyltetracosane
SMILES CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point -38°C
Color Colorless
Density 0.81g/mL at 25°C
pH Neutral
Boiling Point 176°C (0.07 hPa)
Flash Point 218°C (424.4°F)
Odor Odorless
Packaging Glass bottle
Linear Formula [(CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)CH2CH2-]2
Quantity 50 mL
UN Number NONH for all modes of transport
Beilstein 776019
Solubility Information Insoluble in water
Formula Weight 422.81g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material Squalane
Show More Show Less