missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Squalane

Catalog No. 234311500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
234311500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 234311500G Supplier Sigma Aldrich Supplier No. 234311500G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Squalane

Specifications

CAS 111-01-3
Melting Point -38°C (lit.)
Density 0.81 g/mL (at 25°C (literature))
Boiling Point 176°C (0.05 mmHg)
Molecular Formula C30H62
Linear Formula [(CH3)2 CH(CH2)3 CH(CH3)(CH2)3 CH(CH3)CH2CH2-]2
Refractive Index n20/D 1.452 (literature)
MDL Number MFCD00008953
Quantity 500 g
Synonym 2,6,10,15,19,23-Hexamethyltetracosane; Cosbiol; Perhydrosqualene; Robane
Molecular Weight (g/mol) 422.81
Percent Purity 96%
Show More Show Less