Learn More
MilliporeSigma SQPAK™ TOC Quanta Cartridge
Catalog No. SQPAKT0CM1
Change view
Click to view available options
Purification Method: Ion Exchange
Catalog No.
SQPAKT0CM1 Purification Method: Ion Exchange
1 options
Catalog No. SQPAKT0CM1 Supplier MilliporeSigma Supplier No. SQPAKT0CM1
Only null left
Produces ultrapure (Type 1) water with TOC ≤5 ppb. Use with the ultrapure dispensing module of Milli-Q™ SQ 2Series systems
  • Removes organics and ions from pure water.
  • Produce Ultrapure (Type 1) water at 18.2 MΩ·cm resistivity and ≤ 5 ppb TOC.
  • Easy replacement when exhausted thanks to the Twist and Lock mechanism.
  • For use on the ultrapure dispensing module of Milli-Q™ SQ 2Series configurations, including Milli-Q™ SQ 200/200P and SQ 240C/240CV systems
Specifications
Purification Method Ion Exchange
Resin Type Ion Exchange Resin