Learn More
MilliporeSigma SQPAK™ Gard Pack
Description
- Removes free chlorine, colloids and particulates from tap water feed.
- Easy replacement when exhausted thanks to the Twist and Lock mechanism.
- For use on the Milli-Q™ SP 240 Prefilter module.
- Protects reverse osmosis (RO) membranes of Milli-Q™ SR 240/240L/240XL and SQ 240C/240CV systems
Specifications
Specifications
| Feedwater Source | Tap |
| Purification Method | Activated Carbon |