Medical Electronic Systems SQA-V-QWIK-CHECK QC BEADS
Catalog No. NC0343944
QwikCheck Beads is a quality control material for use in automated and manual sperm counting systems. The beads are for in-vitro use only and can be used to assess the accuracy and precision of the laboratory's sperm counting methods.
Catalog No.
NC0343944
Supplier
Medical Electronic SystemsSupplier No.
0200
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
QwikCheck Beads is a quality control material for use in automated and manual sperm counting systems. The beads are for in-vitro use only and can be used to assess the accuracy and precision of the laboratory's sperm counting methods.