missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SQ 22536, 95%

$275.07

Chemical Identifiers

CAS 17318-31-9
Molecular Formula C9H11N5O
Synonym 9-THF-Ade; 6-Amino-9-(tetrahydro-2-furyl)purine
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ67455LB0
View Documents Promotion Details
Thermo Scientific Chemicals
J67455LB0
5 mg
Each for $275.07
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17318-31-9
C9H11N5O
9-THF-Ade; 6-Amino-9-(tetrahydro-2-furyl)purine

Specifications

White to off-white
17318-31-9
205.22
160°C to 161°C
UKHMZCMKHPHFOT-UHFFFAOYNA-N
9-(oxolan-2-yl)-9H-purin-6-amine
95%
-30°C to -10°C
C9H11N5O
9-THF-Ade; 6-Amino-9-(tetrahydro-2-furyl)purine
Solid
NC1=C2N=CN(C3CCCO3)C2=NC=N1
205.22
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only